Please login to your online account to display your discounted pricing

TCR beta Armenian Hamster anti-Mouse, Clone: H57-597, Functional Grade, eBioscience™

Armenian Hamster Monoclonal Antibody, Functional Grade

$100.05 - $361.05

Specifications

Antigen TCR beta
Clone H57-597
Host Species Armenian Hamster
Species Reactivity Mouse
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122973

 
life technologies
LS16596185
500μg Each for $361.05

5014654

 
life technologies
LS16596181
50μg Each for $100.05

5014655

 
life technologies
LS16596182
100μg Each for $201.55
Description & Specifications

Specifications

Antigen TCR beta
Clone H57-597
Host Species Armenian Hamster
Species Reactivity Mouse
Applications Flow Cytometry
Applications Functional Assay
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, no sodium azide
Concentration 1mg/mL

The H57-597 monoclonal antibody reacts with the beta chain of mouse TCR. TCR beta is expressed by thymocytes in a developmentally regulated manner and a majority of peripheral T cells. Crosslinking of the TCR complex with H57-597 induces activation and proliferation of T cells or apoptosis based on assay conditions. H57-597 is used as a phenotypic marker for TCR beta expressing T cells.