Please login to your online account to display your discounted pricing

TCPOBOP, Tocris


Constitutive androstane receptor agonist

Quantity 50mg

Manufacturer: r & d systems  RDS122550

 View more versions of this product

Catalog No. 122550

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 402.06
Quantity 50mg
Molecular Formula C16H8Cl4N2O2
Chemical Name or Material TCPOBOP
Assay Percent Range >98%
CAS 76150-91-9
Synonym 1, 4-Bis(3, 5-Dichloro-2-pyridinyloxy)benzene
Recommended Storage Store at +4°C

Specifications

Chemical Name or Material TCPOBOP
Assay Percent Range >98%
Molecular Formula C16H8Cl4N2O2
Formula Weight 402.06
Synonym 1, 4-Bis(3, 5-Dichloro-2-pyridinyloxy)benzene
Accessories

Store at +4°C