Please login to your online account to display your discounted pricing

TCPOBOP, Tocris

Constitutive androstane receptor agonist

$129.05 - $546.19

Chemical Identifiers

Chemical Name or Material TCPOBOP
Assay Percent Range >98%
Molecular Formula C16H8Cl4N2O2
Formula Weight 402.06
Synonym 1, 4-Bis(3, 5-Dichloro-2-pyridinyloxy)benzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

122510

 
r & d systems
RDS122510
10mg Each for $129.05

122550

 
r & d systems
RDS122550
50mg Each for $546.19
Description & Specifications

Specifications

Formula Weight 402.06
Quantity 10mg
Chemical Name or Material TCPOBOP
Assay Percent Range >98%
Molecular Formula C16H8Cl4N2O2
CAS 76150-91-9
Synonym 1, 4-Bis(3, 5-Dichloro-2-pyridinyloxy)benzene
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C