Please login to your online account to display your discounted pricing

TCN 238, Tocris™

Positive allosteric modulator of mGlu4

$225.77 - $924.23

Chemical Identifiers

Chemical Name or Material TCN 238
Assay Percent Range >98%
Molecular Formula C12H11N3
Formula Weight 197.24
Synonym (E)-4-(2-Phenylethenyl)-2-pyrimidinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

425910

 
r & d systems
425910
10mg Each for $225.77

425950

 
r & d systems
425950
50mg Each for $924.23
Description & Specifications

Specifications

Formula Weight 197.24
Quantity 10mg
Chemical Name or Material TCN 238
Assay Percent Range >98%
Molecular Formula C12H11N3
CAS 125404-04-8
Synonym (E)-4-(2-Phenylethenyl)-2-pyrimidinamine
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C