Please login to your online account to display your discounted pricing

TCN 201, Tocris™

Selective NR1/NR2A receptor antagonist

$246.52 - $1,023.37

Chemical Identifiers

Chemical Name or Material TCN 201
Assay Percent Range >99%
Molecular Formula C21H17ClFN3O4S
Formula Weight 461.89
Synonym 3-Chloro-4-fluoro-N-[4-[[2-(phenylcarbonyl)hydrazino]carbonyl]benzyl]benzenesulfonamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

415450

 
r & d systems
RDS415450
50mg Each for $1,023.37

415410

 
r & d systems
RDS415410
10mg Each for $246.52
Description & Specifications

Specifications

Formula Weight 461.89
Quantity 50mg
Chemical Name or Material TCN 201
Molecular Formula C21H17ClFN3O4S
Synonym 3-Chloro-4-fluoro-N-[4-[[2-(phenylcarbonyl)hydrazino]carbonyl]benzyl]benzenesulfonamide
Recommended Storage Store at +4°C
CAS 852918-02-6
Assay Percent Range >99%
Safety and Handling

Store at +4°C