missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

TCLP Standard, Ricca Chemical

Manufacturer:  Ricca Chemical Company RIRTCLP500

 View more versions of this product

Catalog No. RTCLP-500

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

500mL
Clear light colored
7732-18-5,1327-53-3,7789-02-8,10022-31-8,7439-92-1,7697-37-2
7.21,0.09,0.0022,91.0,0.0029,0.02
Liquid
TCLP Standard
94.0,7.5,0.0022,0.09,0.02,0.0031
Amber Poly

Chemical Identifiers

7732-18-5,7697-37-2,10022-31-8,7789-02-8,1327-53-3,7439-92-1
94.0,7.5,0.09,0.02,0.0031,0.0022
TCLP Standard
91.0,7.21,0.09,0.02,0.0029,0.0022
Liquid

Safety and Handling

ShelfLife : 12 Months