missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
TCLP Standard, Ricca Chemical
Specifications
Chemical Name or Material | TCLP Standard |
---|---|
CAS Min % | 91.0,7.21,0.09,0.02,0.0029,0.0022 |
CAS Max % | 94.0,7.5,0.09,0.02,0.0031,0.0022 |
CAS | 7789-02-8,10022-31-8,1327-53-3,7439-92-1,7697-37-2, 7732-18-5 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
RTCLP-500
|
Ricca Chemical Company
RTCLP500 |
500 mL | Amber Poly Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
RTCLP-100
|
Ricca Chemical Company
RTCLP100 |
100 mL | Amber Poly Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Specifications
TCLP Standard | |
91.0,7.21,0.09,0.02,0.0029,0.0022 | |
94.0,7.5,0.09,0.02,0.0031,0.0022 | |
7789-02-8,10022-31-8,1327-53-3,7439-92-1,7697-37-2, 7732-18-5 |