TCLP Extraction Fluid No. 2, Certified, LabChem™
Manufacturer:  Lab Chem LC259975
 View more versions of this product
Catalog No. LC259975
Specifications
Specifications
TCLP Extraction Fluid No. 2
99.43,0.57
60.05
Certified
Cubitainer™
1.02cSt
2.88 ±0.05
20L
64-19-7,7732-18-5
CH3COOH
Liquid
Passes Test
Soluble in water
0.999g/mL
Colorless
Documents