Please login to your online account to display your discounted pricing

TCLP Extraction Fluid 2, pH 2.88±0.05 at 25°C, for use in EPA 1311 Toxicity Characteristic Leaching Procedure, Ricca Chemical

Chemical Identifiers

Chemical Name or Material TCLP Extraction Fluid 2
CAS 7732-18-5
CAS Min % 97.0
CAS Max % 100.0
CAS 64-19-7
CAS Min % 0.58
CAS Max % 0.61
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

8378-1

 
ricca chemical company
RI83781
Poly natural 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

837825

 
ricca chemical company
RI83782.5
Cubitainer™ 10L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

8378-5

 
ricca chemical company
RI83785
Cubitainer™ 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 4L
CAS 7732-18-5
Chemical Name or Material TCLP Extraction Fluid 2
CAS Min % 97.0
Methods EPA 1311 Toxicity Characteristic Leaching Procedure
CAS 64-19-7
CAS Max % 100.0
Packaging Poly natural
CAS Max % 0.61
CAS Min % 0.58