Please login to your online account to display your discounted pricing

TCLP Extraction Fluid 2 (Certified), Fisher Chemical

Mixture of Acetic Acid and ASTM Type 2 Water

$2,538.08 - $2,538.08

Chemical Identifiers

Chemical Name or Material TCLP Extraction Fluid 2
CAS 64-19-7
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00036152
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

ST200-20

  SDS
fisher scientific company
FLST200-20
PolyPac™ 20L Each for $2,538.08
Description & Specifications

Specifications

Boiling Point 100°C
Melting Point 0°C
Purity Grade Certified Grade
Quantity 20L
Physical Form Liquid
Viscosity [NULL]
Color Clear
pH 2.83 to 2.93
Chemical Name or Material TCLP Extraction Fluid 2
Zinc (Zn) Report ppm
Volatile Background Pass Test
Arsenic (As) 2ppm max.
Barium (Ba) 5ppm max.
Boron (B) Report ppm
Cadmium (Cd) 1ppm max.
Calcium (Ca) Report ppm
Chromium (Cr) 0.5ppm max.
MDL Number MFCD00036152
ChemAlert Storage Symbol Gray
Copper (Cu) Report ppm
Iron (Fe) Report ppm
Manganese (Mn) Report ppm
Magnesium (Mg) Report ppm
Lead (Pb) 0.1ppm max.
Mercury (Hg) 0.01ppm max.
CAS 7732-18-5
Nickel (Ni) Report ppm
Packaging PolyPac™
Aluminum (Al) Report ppm
Selenium (Se) 0.1ppm max.
Semivolatile Background Pass Test
Silver (Ag) 0.1ppm max.
Titanium (Ti) Report ppm
CAS 64-19-7
Safety and Handling