Please login to your online account to display your discounted pricing

TCLP Base/Neutrals Mixture (For EPA TCLP Method 1311)


Quantity 1x1mL
Packaging Ampule

Manufacturer: ultra scientific  TCLP511N1

 View more versions of this product

Catalog No. US-TCLP-511N1

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 1x1mL
Concentration or Composition (by Analyte or Components) Each component at 1000μg/mL in Acetone:
2, 4-Dinitrotoluene
Hexachlorobenzene
Hexachlorobutadiene
1, 4-Dichlorobenzene
Hexachloroethane
Nitrobenzene
Pyridine
Methods For EPA TCLP Method 1311
Packaging Ampule
Chemical Name or Material TCLP Base/Neutrals Mixture

Specifications

Chemical Name or Material TCLP Base/Neutrals Mixture