Please login to your online account to display your discounted pricing

TCLP Base/Neutrals Mixture (For EPA TCLP Method 1311)

$100.51 - $100.51

Chemical Identifiers

Chemical Name or Material TCLP Base/Neutrals Mixture
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

US-TCLP-511N1

 
ultra scientific
TCLP511N1
Ampule 1x1mL Each for $100.51
Description & Specifications

Specifications

Quantity 1x1mL
Concentration or Composition (by Analyte or Components) Each component at 1000μg/mL in Acetone:
2, 4-Dinitrotoluene
Hexachlorobenzene
Hexachlorobutadiene
1, 4-Dichlorobenzene
Hexachloroethane
Nitrobenzene
Pyridine
Methods For EPA TCLP Method 1311
Chemical Name or Material TCLP Base/Neutrals Mixture
Packaging Ampule

Page 313C, Designates A2LA-Certified product