Please login to your online account to display your discounted pricing

TC-P 262, Tocris™

Selective P2X3, P2X2/3 antagonist

$188.60 - $794.96

Chemical Identifiers

Chemical Name or Material TC-P 262
Assay Percent Range >99%
Molecular Formula C14H18N4O
Formula Weight 258.32
Synonym 5-[5-Methyl-2-(1-methylethyl)phenoxy]-2, 4-pyrimidinediamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

438650

 
r & d systems
438650
50mg Each for $794.96

438610

 
r & d systems
438610
10mg Each for $188.60
Description & Specifications

Specifications

Formula Weight 258.32
Quantity 50mg
Chemical Name or Material TC-P 262
Assay Percent Range >99%
Synonym 5-[5-Methyl-2-(1-methylethyl)phenoxy]-2, 4-pyrimidinediamine
Molecular Formula C14H18N4O
CAS 873398-67-5
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature