Please login to your online account to display your discounted pricing

TC-O 9311, Tocris™

Potent and selective GPR139 agonist

$225.77 - $924.23

Chemical Identifiers

Chemical Name or Material TC-O 9311
Assay Percent Range >98%
Molecular Formula C20H19N3O4
Formula Weight 365.38
Synonym 3, 5-Dimethoxybenzoic acid 2-[(1-naphthalenylamino)carbonyl]hydrazide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

425510

 
r & d systems
RDS425510
10mg Each for $225.77

425550

 
r & d systems
RDS425550
50mg Each for $924.23
Description & Specifications

Specifications

Formula Weight 365.38
Quantity 10mg
Chemical Name or Material TC-O 9311
Assay Percent Range >98%
Molecular Formula C20H19N3O4
CAS 444932-31-4
Synonym 3, 5-Dimethoxybenzoic acid 2-[(1-naphthalenylamino)carbonyl]hydrazide
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C