Please login to your online account to display your discounted pricing

TC-N 1752, Tocris™

Selective NaV1.7 blocker

$290.95 - $1,205.80

Chemical Identifiers

Chemical Name or Material TC-N 1752
Assay Percent Range >99%
Molecular Formula C25H27F3N6O3
Formula Weight 516.52
Synonym N-[2-Methyl-3-[[4-[4-[[4-(trifluoromethoxy)phenyl]methoxy]-1-piperidinyl]-1, 3, 5-triazin-2-yl]amino]phenyl]acetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

443510

 
r & d systems
RDS443510
10mg Each for $290.95

443550

 
r & d systems
RDS443550
50mg Each for $1,205.80
Description & Specifications

Specifications

Formula Weight 516.52
Quantity 10mg
Chemical Name or Material TC-N 1752
Assay Percent Range >99%
Molecular Formula C25H27F3N6O3
CAS 1211866-85-1
Synonym N-[2-Methyl-3-[[4-[4-[[4-(trifluoromethoxy)phenyl]methoxy]-1-piperidinyl]-1, 3, 5-triazin-2-yl]amino]phenyl]acetamide
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C