missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

TC LPA5 4, Tocris™

LPA5 receptor antagonist

$250.58 - $1,042.86

Chemical Identifiers

Assay Percent Range >98%
Molecular Formula C23H23ClN2O3
Formula Weight 410.89
Synonym 5-(3-Chloro-4-cyclohexylphenyl)-1-(3-methoxyphenyl)-1H-pyrazole-3-carboxylic acid
Chemical Name or Material TC LPA5 4
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
470810 R & D Systems
RDS470810
10mg Each for $250.58
470850 R & D Systems
RDS470850
50mg Each for $1,042.86
Specifications Description & Specifications

Chemical Identifiers

>98%
410.89
TC LPA5 4
C23H23ClN2O3
5-(3-Chloro-4-cyclohexylphenyl)-1-(3-methoxyphenyl)-1H-pyrazole-3-carboxylic acid

Specifications

10mg
>98%
410.89
TC LPA5 4
1393814-38-4
C23H23ClN2O3
5-(3-Chloro-4-cyclohexylphenyl)-1-(3-methoxyphenyl)-1H-pyrazole-3-carboxylic acid

Safety and Handling

Recommended Storage : Store at +4°C