Please login to your online account to display your discounted pricing

TC-H 106, Tocris™

Class I histone deacetylase inhibitor

$221.01 - $904.70

Chemical Identifiers

Chemical Name or Material TC-H 106
Assay Percent Range >99%
Molecular Formula C20H25N3O2
Formula Weight 339.43
Synonym N1-(2-Aminophenyl)-N7-(4-methylphenyl)heptanediamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

427010

 
r & d systems
RDS427010
10mg Each for $221.01

427050

 
r & d systems
RDS427050
50mg Each for $904.70
Description & Specifications

Specifications

Chemical Name or Material TC-H 106
CAS 937039-45-7
Recommended Storage Store at -20°C
Assay Percent Range >99%
Quantity 10mg
Molecular Formula C20H25N3O2
Formula Weight 339.43
Synonym N1-(2-Aminophenyl)-N7-(4-methylphenyl)heptanediamide
Safety and Handling

Store at -20°C