Please login to your online account to display your discounted pricing

TC-G 1004, Tocris™

Potent and selective A2A antagonist

$266.69 - $1,121.62

Chemical Identifiers

Chemical Name or Material TC-G 1004
Assay Percent Range >98%
Molecular Formula C22H27N7O2
Formula Weight 421.5
Synonym N-[2-(3, 5-Dimethyl-1H-pyrazol-1-yl)-6-[6-(4-methoxy-1-piperidinyl)-2-pyridinyl]-4-pyrimidinyl]acetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

440710

 
r & d systems
440710
10mg Each for $266.69

440750

 
r & d systems
440750
50mg Each for $1,121.62
Description & Specifications

Specifications

Formula Weight 421.5
Quantity 10mg
Assay Percent Range >98%
Synonym N-[2-(3, 5-Dimethyl-1H-pyrazol-1-yl)-6-[6-(4-methoxy-1-piperidinyl)-2-pyridinyl]-4-pyrimidinyl]acetamide
CAS 1061747-72-5
Chemical Name or Material TC-G 1004
Molecular Formula C22H27N7O2
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C