Please login to your online account to display your discounted pricing

TC ASK 10, Tocris™

Potent and selective ASK1 inhibitor; orally bioavailable

$284.81 - $1,180.32

Chemical Identifiers

Chemical Name or Material TC ASK 10
Assay Percent Range >98%
Molecular Formula C21H21N5O·2HCl
Formula Weight 432.35
Synonym 4-(1, 1-Dimethylethyl)-N-[6-(1H-imidazol-1-yl)imidazo[1, 2-a]pyridin-2-yl]benzamide dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

482510

 
r & d systems
RDS482510
10mg Each for $284.81

482550

 
r & d systems
RDS482550
50mg Each for $1,180.32
Description & Specifications

Specifications

Formula Weight 432.35
Quantity 10mg
Synonym 4-(1, 1-Dimethylethyl)-N-[6-(1H-imidazol-1-yl)imidazo[1, 2-a]pyridin-2-yl]benzamide dihydrochloride
Recommended Storage Desiccate at Room Temperature
Chemical Name or Material TC ASK 10
Assay Percent Range >98%
CAS 1005775-56-3
Molecular Formula C21H21N5O·2HCl
Safety and Handling

Desiccate at Room Temperature