Please login to your online account to display your discounted pricing

TC-A 2317 hydrochloride, Tocris™

Potent, selective Aurora kinase A inhibitor

$290.95 - $1,205.80

Chemical Identifiers

Chemical Name or Material TC-A 2317 hydrochloride
Assay Percent Range >98%
Molecular Formula C19H28N6O·HCl
Formula Weight 392.93
Synonym 2-[(5-Hydroxy-1, 5-dimethylhexyl)amino]-4-methyl-6-[(5-methyl-1H-pyrazol-3-yl)amino]-3-pyridinecarbonitrile hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

406650

 
r & d systems
RDS406650
50mg Each for $1,205.80

406610

 
r & d systems
RDS406610
10mg Each for $290.95
Description & Specifications

Specifications

Formula Weight 392.93
Quantity 50mg
Chemical Name or Material TC-A 2317 hydrochloride
Assay Percent Range >98%
Molecular Formula C19H28N6O·HCl
CAS 1245907-03-2
Synonym 2-[(5-Hydroxy-1, 5-dimethylhexyl)amino]-4-methyl-6-[(5-methyl-1H-pyrazol-3-yl)amino]-3-pyridinecarbonitrile hydrochloride
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C