Please login to your online account to display your discounted pricing

TC 2559 difumarate, Tocris

Selective partial agonist at α4β2 receptors

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material TC 2559 difumarate
Assay Percent Range >98%
Molecular Formula C12H18N2O·2C4H4O4
Formula Weight 438.43
Synonym 4-(5-ethoxy-3-pyridinyl)-N-methyl-(3E)-3-buten-1-amine difumarate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

273710

 
r & d systems
RDS273710
10mg Each for $239.63

273750

 
r & d systems
RDS273750
50mg Each for $976.15
Description & Specifications

Specifications

Formula Weight 438.43
Quantity 10mg
Chemical Name or Material TC 2559 difumarate
CAS 212332-35-9
Assay Percent Range >98%
Molecular Formula C12H18N2O·2C4H4O4
Synonym 4-(5-ethoxy-3-pyridinyl)-N-methyl-(3E)-3-buten-1-amine difumarate
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature