Please login to your online account to display your discounted pricing

TC 1698 dihydrochloride, Tocris

α7-selective agonist

$214.12 - $895.76

Chemical Identifiers

Chemical Name or Material TC 1698 dihydrochloride
Assay Percent Range >98%
Molecular Formula C13H18N2·2HCl
Formula Weight 275.22
Synonym 2-(3-Pyridinyl)-1-azabicyclo[3.2.2]nonane dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

251850

 
r & d systems
RDS251850
50mg Each for $895.76

251810

 
r & d systems
RDS251810
10mg Each for $214.12
Description & Specifications

Specifications

Formula Weight 275.22
Quantity 50mg
Chemical Name or Material TC 1698 dihydrochloride
Assay Percent Range >98%
Molecular Formula C13H18N2·2HCl
CAS 787587-06-8
Synonym 2-(3-Pyridinyl)-1-azabicyclo[3.2.2]nonane dihydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature