Please login to your online account to display your discounted pricing

TC 1, Tocris

High affinity σ1 ligand

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material TC 1
Assay Percent Range >99%
Molecular Formula C19H20FNO
Formula Weight 297.37
Synonym (N-(3'-Fluorophenyl)ethyl-4-azahexacyclo[5.4.1.02,6.03,10.05,9.08,11]dodecan-3-ol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

266910

 
r & d systems
266910
10mg Each for $252.39

266950

 
r & d systems
266950
50mg Each for $1,050.42
Description & Specifications

Specifications

Formula Weight 297.37
Quantity 10mg
CAS 362512-81-0
Chemical Name or Material TC 1
Synonym (N-(3'-Fluorophenyl)ethyl-4-azahexacyclo[5.4.1.02,6.03,10.05,9.08,11]dodecan-3-ol
Assay Percent Range >99%
Molecular Formula C19H20FNO
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature