General Data Healthcare™ TMD™-7 Tissue Marking Dyes Refills DFS Item
Refills for the TMD-7 Tissue Marking Dyes 7 Color Kit
Manufacturer:  General Data Healthcare™ TMDG*
 View more versions of this product
Catalog No. 15-183-45
Specifications
Specifications
Liquid
8 oz.
Tissue Marking Dye™
2-Propanol 7%, Ammonium Hydroxide 2%, Formaldehyde 0.05%, N,N-Dimethylaminoethanol 2%, Water 75%
Green
4 Case
Documents