General Data Healthcare™ TFM™ Colored Tissue Freezing Medium DFS Item
Variety of colors to easily differentiate multiple specimens during sectioning and staining
Manufacturer:  General Data Healthcare™ TFMB
 View more versions of this product
Catalog No. 15-183-37
Description
Description
  • Unique formula reduces curling, allowing easier pickup of flat serial sections
  • Reduced water content minimizes freeze fractures
Order Info
Supplied in 4 oz. (118mL) plastic squeeze bottles. Complete set includes one of all five medium colors in a wooden rack.
Specifications
Specifications
212°C
Embedding matrix for frozen sections
Tissue Freezing Medium™
0°C
Blue
118mL
Documents