Please login to your online account to display your discounted pricing

TBS Tablets, Calbiochem™, EMD Millipore

Dissolving one tablet in 500mL of deionized H2O yields 150mM NaCl, 50mM Tris-HCl buffer

$61.00 - $61.00

Chemical Identifiers

Chemical Name or Material TBS Tablets
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

5247501EA

 
emd millipore corporation
5247501EA
Each for $61.00
Description & Specifications

Specifications

Quantity 1 strip (10 tablets)
Chemical Name or Material TBS Tablets
Synonym Tris Buffered Saline 10 Tablets
Solubility Information Soluble in water
Recommended Storage 15°C to 30°C (Do not freeze).
Safety and Handling

15°C to 30°C (Do not freeze).