Please login to your online account to display your discounted pricing

TBE Buffer, 10X Powder Pack, ULTROL™ Grade, Calbiochem™, EMD Millipore

Each pack provides an adequate amount of material to make 1L of 10X concentrate that can be diluted to a 1X solution containing 89mM Tris, 89mM boric acid, 2mM EDTA

$81.70 - $81.70

Chemical Identifiers

Analyte or Component Names Tris, EDTA, Boric acid
Chemical Name or Material TBE Buffer
Concentration or Composition (by Analyte or Components) 1X solution containing 89mM Tris, 89mM boric acid, 2mM EDTA
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

5747962PACK

 
emd millipore corporation
5747962PACK
Each for $81.70
Description & Specifications

Specifications

Quantity 2 Pk.
Chemical Name or Material TBE Buffer
Analyte or Component Names Tris, EDTA, Boric acid
DNase None Detected
Synonym 10X Tris-Borate-EDTA Buffer
Concentration or Composition (by Analyte or Components) 1X solution containing 89mM Tris, 89mM boric acid, 2mM EDTA
Solubility Information Easily soluble in cold water
Protease None Detected
Purity Molecular Biology Grade
Recommended Storage 15°C to 30°C (Do not freeze); Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).
RNase None Detected
Safety and Handling

15°C to 30°C (Do not freeze); Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).