Please login to your online account to display your discounted pricing

TBE Buffer, 10X, Molecular Biology Grade, Calbiochem™, EMD Millipore

A 10X concentrate that can be diluted to a 1X solution containing 89mM Tris, 89mM boric acid, 2mM EDTA

$57.94 - $57.94

Chemical Identifiers

Analyte or Component Names Tris, EDTA, Boric acid
Chemical Name or Material TBE Buffer
Concentration or Composition (by Analyte or Components) 1X solution contains 89mM Tris, 89mM boric acid, 2mM EDTA
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

5747951L

 
emd millipore corporation
5747951L
Each for $57.94
Description & Specifications

Specifications

Quantity 1L
Purity Molecular Biology Grade
Analyte or Component Names Tris, EDTA, Boric acid
Protease None Detected
RNase None Detected
Chemical Name or Material TBE Buffer
DNase None Detected
Concentration or Composition (by Analyte or Components) 1X solution contains 89mM Tris, 89mM boric acid, 2mM EDTA
Recommended Storage 15°C to 30°C (Do not freeze).
Synonym Tris-Borate-EDTA Buffer
Safety and Handling

15°C to 30°C (Do not freeze).