Please login to your online account to display your discounted pricing

Alfa Aesar™ TB Solution-II

For transformation of competent cells

Chemical Identifiers

Chemical Name or Material TB Solution-II
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ60028EQE

  SDS
alfa aesar™
J60028EQE
125mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material TB Solution-II
Physical Form Liquid
Color Colorless
Quantity 125mL
Recommended Storage 2° to 8°C
Safety and Handling

2° to 8°C