Please login to your online account to display your discounted pricing

TB 21007, Tocris

α5-selective GABAA inverse agonist

$212.74 - $886.68

Chemical Identifiers

Chemical Name or Material TB 21007
Assay Percent Range >99%
Molecular Formula C15H17NO2S3
Formula Weight 339.5
Synonym 6, 7-Dihydro-3-[(2-hydroxyethyl)thio]-6, 6-dimethyl-1-(2-thiazolyl)-benzo[c]thiophen-4(5H)-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

29-051-0

 
r & d systems
RDS290210
10mg Each for $212.74

290550

 
r & d systems
RDS290550
50mg Each for $886.68
Description & Specifications

Specifications

Formula Weight 339.5
Quantity 10mg
Chemical Name or Material TB 21007
CAS 207306-50-1
Synonym 6, 7-Dihydro-3-[(2-hydroxyethyl)thio]-6, 6-dimethyl-1-(2-thiazolyl)-benzo[c]thiophen-4(5H)-one
Recommended Storage Store at +4°C
Molecular Formula C15H17NO2S3
Assay Percent Range >99%
Safety and Handling

Store at +4°C