missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tazarotene, TCI America™

Manufacturer:  TCI America T310810MG

 View more versions of this product

Catalog No. T310810MG



Specifications

Specifications

Tazarotene
105°C
118292-40-3
MFCD00867628
OGQICQVSFDPSEI-UHFFFAOYSA-N
ethyl 6-[2-(4,4-dimethyl-2,3-dihydrothiochromen-6-yl)ethynyl]pyridine-3-carboxylate
5381
351.46
≥98.0% (GC)
Yellow
10mg
C21H21NO2S
tazarotene, tazorac, zorac, avage, ethyl 6-4,4-dimethylthiochroman-6-yl ethynyl nicotinate, fabior, tazarotene usan:inn, tazarotene avage, tazaroteno, tazarotenum
CCOC(=O)C1=CN=C(C=C1)C#CC2=CC3=C(C=C2)SCCC3(C)C
351.464
CHEBI:32184
Crystal-Powder at 20°C

Chemical Identifiers

118292-40-3
351.464
OGQICQVSFDPSEI-UHFFFAOYSA-N
5381
ethyl 6-[2-(4,4-dimethyl-2,3-dihydrothiochromen-6-yl)ethynyl]pyridine-3-carboxylate
C21H21NO2S
MFCD00867628
tazarotene, tazorac, zorac, avage, ethyl 6-4,4-dimethylthiochroman-6-yl ethynyl nicotinate, fabior, tazarotene usan:inn, tazarotene avage, tazaroteno, tazarotenum
CHEBI:32184
CCOC(=O)C1=CN=C(C=C1)C#CC2=CC3=C(C=C2)SCCC3(C)C
Safety and Handling

Safety and Handling

RTECSNumber : US5675100

TSCA : No

Recommended Storage : Refrigerator

Documents