missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Tau Ab-3, Rabbit Polyclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Tau Ab-3, Rabbit Polyclonal Antibody.

$551.29 - $1,028.00

Specifications

Antigen TAU Ab-3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
RB1429A View Documents Thermo Scientific™
RB1429A
1mL Each for $1,028.00
 
RB1429A1 View Documents Thermo Scientific™
RB1429A1
500μL Each for $708.67
 
RB1429R7 View Documents Thermo Scientific™
RB1429R7
7mL Each for $551.29
 
Description

Description

Provide accurate, reproducible results with the Thermo Scientific™ Tau Ab-3, Rabbit Polyclonal Antibody.

Tau proteins are microtubule-associated proteins (MAPs). Alternate splicing of tau mRNA and differential phosphorylation contribute to the heterogeneity of tau.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not known.

Epitope: C-terminal

Immunogen: Recombinant protein encoding C-terminal part of human tau

Molecular Weight: 45-68kDa

Positive Control: Alzheimer's disease brain tissue

Cellular Localization: Cytoplasmic Ab-3 stains the human neurofibrillary tangles, neuropil threads, and neuritic plaques associated with Alzheimer's disease. It also stains astrocytes and ribosomes.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

TAU Ab-3
Polyclonal
Rabbit
Neuroscience
Recombinant protein encoding C-terminal part of human tau
Immunohistochemistry (Paraffin)
Unconjugated
RUO
Primary
IgG