Please login to your online account to display your discounted pricing

TAS 301, Tocris™

Inhibitor of smooth muscle cell migration and proliferation

$177.37 - $746.48

Chemical Identifiers

Chemical Name or Material TAS 301
Assay Percent Range >99%
Molecular Formula C23H19NO3
Formula Weight 357.4
Synonym 3-[Bis(4-methoxyphenyl)methylene]-1, 3-dihydro-2H-indol-2-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

452150

 
r & d systems
RDS452150
50mg Each for $746.48

452110

 
r & d systems
RDS452110
10mg Each for $177.37
Description & Specifications

Specifications

Formula Weight 357.4
Quantity 50mg
Chemical Name or Material TAS 301
Assay Percent Range >99%
Molecular Formula C23H19NO3
CAS 193620-69-8
Synonym 3-[Bis(4-methoxyphenyl)methylene]-1, 3-dihydro-2H-indol-2-one
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C