missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tartaric Acid AR Granular, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 87-69-4
Physical Form Granular
Molecular Formula HOCO(CHOH)2COOH
Formula Weight 150.09
Chemical Name or Material Tartaric Acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK2312500 Avantor Performance
231204*
POLYSTORMOR 500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

87-69-4
HOCO(CHOH)2COOH
Tartaric Acid
Granular
150.09

Specifications

150.09
87-69-4
Granular
Tartaric Acid
500g
HOCO(CHOH)2COOH
POLYSTORMOR