Please login to your online account to display your discounted pricing

Tartaric Acid, ACS Grade, ≥ 99.0%, LabChem™

$139.09 - $139.09

Chemical Identifiers

Chemical Name or Material Tartaric Acid
CAS 87-69-4
CAS Max % 100
Purity Grade ACS Grade
Linear Formula C4H6O6
Formula Weight 150.09
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC259401

 
lab chem
LC259401
Poly Bottle 500g Each for $139.09
Description & Specifications

Specifications

Formula Weight 150.09
Assay ≥99.0%
Recommended Storage Room Temperature
CAS Max % 100
Health Hazard 2 GHS H Statement
Causes serious eye damage.
Harmful to aquatic life.
Quantity 500g
Health Hazard 3 GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Purity Grade ACS Grade
Flash Point 150°C
Chemical Name or Material Tartaric Acid
Solubility Soluble in water
Physical Form Powder
CAS 87-69-4
EINECS Number 201-766-0
Health Hazard 1 Danger
Linear Formula C4H6O6
Decomposition Information Carbon monoxide; Carbon dioxide
Identification Passes Test
Color White
Packaging Poly Bottle
Safety and Handling

Danger

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
Causes serious eye damage.
Harmful to aquatic life.

Room Temperature