Please login to your online account to display your discounted pricing

Tartaric Acid, 2% (w/v) Aqueous Solution, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Tartaric Acid
CAS 7732-18-5
CAS Min % 96.0
CAS Max % 100.0
CAS 87-69-4
CAS Min % 1.92
CAS Max % 2.0
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

8370-32

 
ricca chemical company
RI837032
Poly natural 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

8370-16

 
ricca chemical company
RI837016
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Clear colorless to slightly yellow
Physical Form Liquid
Quantity 1L
CAS 7732-18-5
Chemical Name or Material Tartaric Acid
Concentration or Composition Notes 2% (w/v)
Solvent or Matrix Aqueous Solution
CAS Min % 96.0
CAS Max % 100.0
CAS 87-69-4
Packaging Poly natural
CAS Min % 1.92
CAS Max % 2.0