Please login to your online account to display your discounted pricing

Tartaric Acid, 2% (w/v) Aqueous Solution, Certified APHA Reagent, Ricca Chemical


Quantity 20L
Packaging Cubitainer

Manufacturer: ricca chemical company  RIR837000020F

 View more versions of this product

Catalog No. R837000020F

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Tartaric Acid
Name Note 2% (w/v) Aqueous Solution
CAS 7732-18-5
CAS 87-69-4
Physical Form Liquid
Odor Odorless
Packaging Cubitainer
Solubility Solubility in Water: Infinite
Solvent or Matrix Aqueous Solution
Color Clear colorless to slightly yellow
Quantity 20L

Specifications

Chemical Name or Material Tartaric Acid
Name Note 2% (w/v) Aqueous Solution
CAS 7732-18-5
CAS 87-69-4
Physical Form Liquid