Please login to your online account to display your discounted pricing

Tartaric Acid, 2% (w/v) Aqueous Solution, Certified APHA Reagent, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Tartaric Acid
Name Note 2% (w/v) Aqueous Solution
CAS 7732-18-5
CAS 87-69-4
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

R837000020F

 
ricca chemical company
RIR837000020F
Cubitainer 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R83700004A

 
ricca chemical company
RIR83700004A
Poly natural 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless to slightly yellow
Quantity 20L
Chemical Name or Material Tartaric Acid
Solubility Solubility in Water: Infinite
CAS 7732-18-5
Odor Odorless
Solvent or Matrix Aqueous Solution
CAS 87-69-4
Packaging Cubitainer
Name Note 2% (w/v) Aqueous Solution