Applied Biosystems™ TaqMan™ SARS-CoV-2 Pooling Assay Kit DFS Item
The Applied Biosystems TaqMan SARS-CoV-2 Pooling Assay is a fast research solution for the real-time PCR detection of RNA from the SARS-CoV-2 virus that enables labs to pool up to five samples in a single reaction.
Manufacturer:  Applied Biosystems™ A50790
Catalog No. A50790
Description
Description
Features of the TaqMan SARS-CoV-2 Pooling Assay include:
  • Targets spike (S) protein and nucleocapsid (N) protein regions exhibiting lower risk for mutation
  • May be used to quickly analyze samples in under four hours with one instrument. If a pool returns a positive result, the individual samples used to create the pool should be tested individually. Deconvolution of an individual sample from a pool that has SARS-CoV-2 nucleic acid detected would take less than four hours.
  • Has been tested on the following instruments: QuantStudio 5 Real-Time PCR System (96-well, 0.2 mL), QuantStudio 7 Real-Time PCR System (384 well, 0.2 mL), and associated Design and Analysis 2.5 software
The TaqMan SARS-CoV-2 Pooling Assay kit includes the following components:
  • SARS-CoV-2 real-time PCR pooling assay multiplex—multiplexed assays that contain three primer/probe sets specific to different SARS-CoV-2 genomic regions and primers/probes for bacteriophage MS2
  • MS2 phage control—internal process control for nucleic acid extraction
Required materials not supplied
  • TaqMan SARS CoV-2 Control (Cat. No. 956118)—RNA control that contains targets specific to the SARS-CoV-2 genomic regions targeted by the assays
  • TaqMan SARS CoV-2 Control Dilution Buffer (Cat. No. A49889)—dilution buffer for the IVT RNA control
Specifications
Specifications
• SARS CoV-2 Pooling multiplex Assay, 300 µL, store at -10°C to -30°C
• TaqMan MS2 phage control, 2 x 1,000 µL, store at -10°C to -30°C
SARS CoV-2 Pathogen Research Solutions
200
Human
Primer-Probe Detection
TaqMan
Dry Ice
severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2)
SDS
Documents
Product Certifications
Promotions
Promotions
For Resarch Use Only.