missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Taq DNA Polymerase w/o Detergent

Contains no detergent or BSA. MP Biomedicals Taq DNA Polymerase w/o Detergent is a specialized, thermostable enzyme. The incubation buffer (XD buffer) allows optimal results for researchers working with vegetal or beef material or using further dHPLC.

$290.19 - $2,151.71

Specifications

For Use With (Application) Taq DNA Polymerase is a thermostable DNA polymerase that is used for the DNA polymerase chain reaction (PCR) in order to amplify DNA sequences. Other application include RT-PCR, DNA sequencing and DNA labeling.
Storage Requirements Freeze at −20°C
Formulation Liquid
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MP1EPTQX025
View Documents Promotion Details
MP Biomedicals™
11EPTQX025
250 Units
Each for $290.19
 
MP1EPTQX925
View Documents Promotion Details
MP Biomedicals™
11EPTQX925
10 × 250 units
Each for $2,151.71
 
Description

Description

Contains a specialized, thermostable enzyme isolated from the thermophilic bacterium Thermus aquaticus. The recombinant form of this enzyme is expressed in E. coli.

  • Catalyzes oligonucleotide primer-driven, DNA-template-dependent incorporation of dNTPs into complimentary DNA strands
  • Displays both 5' to 3' polymerase and exonuclease activities
  • Used for the DNA polymerase chain reaction (PCR) in order to amplify DNA sequences
  • Other application include RT-PCR, DNA sequencing and DNA labeling
Specifications

Specifications

Taq DNA Polymerase is a thermostable DNA polymerase that is used for the DNA polymerase chain reaction (PCR) in order to amplify DNA sequences. Other application include RT-PCR, DNA sequencing and DNA labeling.
Freeze at −20°C
Liquid
SDS
Product Certifications