MP Biomedicals™ Taq DNA Polymerase w/o Detergent
Contains no detergent or BSA. MP Biomedicals Taq DNA Polymerase w/o Detergent is a specialized, thermostable enzyme. The incubation buffer (XD buffer) allows optimal results for researchers working with vegetal or beef material or using further dHPLC.
Manufacturer:  MP Biomedicals™ 11EPTQX025
 View more versions of this product
Catalog No. MP1EPTQX025
Description
Description
Contains a specialized, thermostable enzyme isolated from the thermophilic bacterium Thermus aquaticus. The recombinant form of this enzyme is expressed in E. coli.
  • Catalyzes oligonucleotide primer-driven, DNA-template-dependent incorporation of dNTPs into complimentary DNA strands
  • Displays both 5' to 3' polymerase and exonuclease activities
  • Used for the DNA polymerase chain reaction (PCR) in order to amplify DNA sequences
  • Other application include RT-PCR, DNA sequencing and DNA labeling
Specifications
Specifications
Taq DNA Polymerase is a thermostable DNA polymerase that is used for the DNA polymerase chain reaction (PCR) in order to amplify DNA sequences. Other application include RT-PCR, DNA sequencing and DNA labeling.
Liquid
Freeze at −20°C
250 Units
Documents