Thermo Scientific™ Taq DNA Polymerase, Native
Optimize routine PCR with this highly thermostable, native DNA polymerase.
$231.00 - $231.00
Specifications
Concentration 5U/µL
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Units Price Quantity  
Catalog Number Mfr. No. Units Price Quantity  
FEREP0072 Thermo Scientific™
EP0072
300U Each for $231.00
Specifications
Description
Native Taq DNA Polymerase is a highly thermostable DNA polymerase of the thermophilic bacterium Thermus aquaticus. The enzyme catalyzes 5'→3' synthesis of DNA, has no detectable 3'→5' exonuclease (proofreading) activity and possesses low 5'→3' exonuclease activity. In addition, the polymerase exhibits deoxynucleotidyl transferase activity, which frequently results in the addition of extra adenines at the 3'-end of PCR products. Native Taq DNA Polymerase is preferred for amplification of bacterial DNA sequences homologous to those found in E. coli.
Taq DNA Polymerase (native, with BSA) is supplied with BSA as a stabilizing agent. This version of Taq DNA Polymerase is often the best choice when amplifying DNA samples of lower purity, e.g. genomic DNA from mouse tail.
Highlights
  • Thermostable -half life is more than 40 min at 95°C.
  • Generates PCR products with 3'-dA overhangs.
  • Supplied with two buffers -10X Taq Buffer with KCl and 10X Taq Buffer with (NH4)2SO4. The latter allows for PCR at wide range of magnesium concentrations and decreases unspecific priming.
  • Incorporates modified nucleotides (e.g., biotin-, digoxigenin-, fluorescently-labeled nucleotides).
Description & Specifications
Specifications
5U/µL