missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Taq DNA Polymerase Kts

Thermo Scientific™ Taq DNA Polymerase Kits are ideal for routine PCR applications requiring consistent and reliable Taq performance.

$1,943.43 - $1,943.43

Specifications

Concentration 5U/µL
For Use With (Application) Amplifying fragments up to 5kb; PCR; RT-PCR; Sequencing; Genotyping; Library Construction; Cloning; Pathogen Detection
Storage Requirements Store at -20°C. Avoid freeze/thaw cycles
View More Specs

Disclaimers:

Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser's own internal research. No other patent rights are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.



Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Quantity Price Quantity  
Catalog Number Mfr. No. Product Type Quantity Price Quantity  
AB-0192B Thermo Scientific™
AB0192B
Taq DNA Polymerase Enzyme Kit 4000 x 25µL Rxns Each for $1,943.43
Specifications

Description

Thermo Scientific™ Taq DNA Polymerase Kits are ideal for routine PCR applications requiring consistent and reliable Taq performance.

Recommended for: Routine PCR
Description & Specifications

Specifications

5U/µL
Amplifying fragments up to 5kb; PCR; RT-PCR; Sequencing; Genotyping; Library Construction; Cloning; Pathogen Detection
Store at -20°C. Avoid freeze/thaw cycles