Please login to your online account to display your discounted pricing

Thermo Scientific™ Taq DNA Polymerase Kts


Thermo Scientific™ Taq DNA Polymerase Kits are ideal for routine PCR applications requiring consistent and reliable Taq performance.

Manufacturer: thermo scientific™  AB0192B

 View more versions of this product

 Disclaimers

Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser's own internal research. No other patent rights are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.

Catalog No. AB-0192B

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Amplifying fragments up to 5kb; PCR; RT-PCR; Sequencing; Genotyping; Library Construction; Cloning; Pathogen Detection
Product Type Taq DNA Polymerase Enzyme Kit
Quantity 4000 x 25µL Rxns
Storage Requirements Store at -20°C. Avoid freeze/thaw cycles
Concentration 5U/µL
Item Description AB-0192/B
Product Name Taq DNA Polymerase Enzyme Kit
Concentration 5U/µL

Thermo Scientific™ Taq DNA Polymerase Kits are ideal for routine PCR applications requiring consistent and reliable Taq performance.

 Recommended for: 

Routine PCR