Please login to your online account to display your discounted pricing

LaMotte™ Tapwater Tour


Analyze the quality of tap water at home or in the classroom.

Manufacturer: lamotte™  3608

 View more versions of this product

Catalog No. S25705

  • / Each
 

Description & Specifications

Specifications

Includes Lecture materials, illustrated handouts, teacher tips, test procedures and wrap up games, five units
Sufficient For 50 students
Grade 4 to 6
For Use With (Application) Analyze the quality of tap water at home or in the classroom
Product Type Tap Water Tour Kit