Please login to your online account to display your discounted pricing

TAPS, ULTROL™ Grade, Calbiochem™, EMD Millipore

A zwitterionic buffer useful in the pH range of 7.7 to 9.1

$475.00 - $475.00

Chemical Identifiers

Assay Percent Range >98%
Formula Weight 243.2
Buffers zwitterionic buffer
CAS 29915-38-6
Chemical Name or Material TAPS
Molecular Formula C7H17NO6S
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

3946751KG

 
emd millipore corporation
3946751KG
Each for $475.00
Description & Specifications

Specifications

Formula Weight 243.2
Quantity 1kg
Chemical Name or Material TAPS
Buffers zwitterionic buffer
Purity Grade Notes by titration
Assay Percent Range >98%
Molecular Formula C7H17NO6S
CAS 29915-38-6
Synonym 3-{[tris(Hydroxymethyl)methyl]amino}propanesulfonic Acid
Solubility Information Easily soluble in cold water
Purity ULTROL™ Grade
Recommended Storage 15°C to 30°C (Do not freeze); Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).
Safety and Handling

15°C to 30°C (Do not freeze); Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).