Invitrogen™ TAOK3 (JIK) Recombinant Human Protein DFS Item
TAOK3 (JIK) is a serine/threonine kinase that is a member of the GCK-like subfamily of the STE20 kinase family.
Manufacturer:  Invitrogen™ LSPV4069
 View more versions of this product
Catalog No. PV4069
Description
Description
JIK is ubiquitously expressed in human tissues, inhibits JNK and has been implicated in cancer.
Specifications
Specifications
NP_057365
Frozen
134.9kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
STE20 Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
TAOK3 (JIK)
Baculovirus
Human
GST Tag
Documents