missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
TAOK3 (JIK) Recombinant Human Protein
TAOK3 (JIK) Recombinant Human Protein
Supplier: Thermo Fisher Scientific PV3652
Description
TAOK3 (JIK) is a serine⁄threonine kinase that is a member of the GCK-like subfamily of the STE20 kinase family. JIK is ubiquitously expressed in human tissues, inhibits JNK and has been implicated in cancer.Specifications
NP_057365 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
134.9kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
TAOK3 (JIK) | |
TAOK3 was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 100μM ATP, 667μg/mL myelin basic protein (MBP) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
TAOK3 (JIK) Recombinant Human Protein | |
Human | |
Baculovirus | |
Serine/threonine kinase |
0.26mg/mL | |
3552, 51347 | |
TAOK3 (JIK) Recombinant Human Protein | |
–80°C | |
1 Each | |
TAOK3 (JIK) Recombinant Human Protein | |
STE20 | |
47nmoles of phosphate transferred to myelin basic protein (MBP) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 3.33μg/mL | |
GST Tag | |
Kinase and inhibitors | |
65% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.