missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ TAOK2 (TAO1) Recombinant Human Protein
TAOK2 (TAO1) is a serine/threonine kinase.
Manufacturer: Invitrogen™ LSPV4068
Description
TAOK2 (TAO1) may be an important regulator of the stress-responsive p38 MAP kinase pathway.
Specifications
NP_004774 | |
Frozen | |
63.1kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Catalytic | |
Kinase Assay | |
STE20 Kinase Family | |
Yes |
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
1mg | |
RUO - research use only | |
TAOK2 (TAO1) | |
Baculovirus | |
Human | |
GST Tag |