TAOK2 (TAO1) Recombinant Human Protein DFS Item
TAOK2 (TAO1) Recombinant Human Protein
Manufacturer:  Life Technologies LSPV3760
Catalog No. PV3760
Description
Description
TAOK2 (TAO1) is a serine⁄threonine kinase that may be an important regulator of the stress-responsive p38 MAP kinase pathway.
Specifications
Specifications
NP_004774
Pharma and biopharma, protein assays and analysis, target and lead identification and validation
63.1kDa
85% by SDS-PAGE
–80°C
1 Each
TAOK2 (TAO1)
TAOK2 (TAO1) was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 667μg/mL myelin basic protein (MBP) and trace [32P]-γ-ATP for 10 minutes at 30°C
Kinase and inhibitors
STE20
TAOK2 (TAO1) Recombinant Human Protein
Human
0.48mg/mL
6532, 9344
TAOK2 (TAO1) Recombinant Human Protein
10μg
For research use only. Not for use in diagnostic procedures
Serine/threonine kinase
TAOK2 (TAO1) Recombinant Human Protein
Kinase assay
Baculovirus
Amino acids 1-314
340nmoles of phosphate transferred to myelin basic protein (MBP) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 1.67μg/mL
GST Tag
Documents
For Research Use Only. Not for use in diagnostic procedures.